Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx9
Entrez Gene ID
66616
Gene Name
sorting nexin 9
Gene Aliases
2700073N08Rik, SDP1, SH3PX1
Chromosome Location
Chr.17: 5841380 - 5930711 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025664.5 NP_079940.2 9 - 10 1100 73
GenBank mRNA AK012520.1 - 4 - 5 392 73
AK139802.1 - 9 - 10 1134 73
AK149739.1 - 9 - 10 1114 73
AK149849.1 - 9 - 10 1154 73
AK155830.1 - - 1135 73
AK157517.1 - 9 - 10 1116 73
AK170339.1 - - 1116 73
BC014814.1 - 9 - 10 1100 73

View Full Product Details