Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx9
Entrez Gene ID
66616
Gene Name
sorting nexin 9
Gene Aliases
2700073N08Rik, SDP1, SH3PX1
Chromosome Location
Chr.17: 5841380 - 5930711 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025664.5 NP_079940.2 14 - 15 1601 65
GenBank mRNA AK012520.1 - 9 - 10 893 65
AK139802.1 - 14 - 15 1635 65
AK149739.1 - 14 - 15 1614 65
AK149849.1 - 14 - 15 1655 65
AK155830.1 - - 1636 65
AK157517.1 - 14 - 15 1617 65
AK170339.1 - - 1617 65
BC014814.1 - 14 - 15 1601 65

View Full Product Details