Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stt3a
Entrez Gene ID
16430
Gene Name
STT3, subunit of the oligosaccharyltransferase complex, homolog A (S. cerevisiae)
Gene Aliases
AA408947, BB081708, Itm1
Chromosome Location
Chr.9: 36729345 - 36767674 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008408.4 NP_032434.3 15 - 16 1837 86
GenBank mRNA AK030363.1 - 15 - 16 1901 86
AK050050.1 - 1 - 2 105 86
AK077877.1 - 15 - 16 1860 86
AK081547.1 - 15 - 16 1894 86
AK087470.1 - 15 - 16 1884 86
AK150640.1 - 15 - 16 1882 86
AK151541.1 - 15 - 16 1893 86
AK151791.1 - 15 - 16 1893 86
AK152242.1 - 15 - 16 1882 86
AK153840.1 - 15 - 16 1838 86
AK153886.1 - 15 - 16 1938 86
AK160037.1 - 15 - 16 1876 86
AK160607.1 - 15 - 16 1838 86
AK168505.1 - 15 - 16 1883 86
AK168937.1 - 15 - 16 1882 86
AK169669.1 - 15 - 16 1861 86
AX799090.1 - 15 - 16 1880 86
BC028897.1 - 14 - 15 1786 86
BC037612.1 - 15 - 16 1876 86
BC085313.1 - 15 - 16 1866 86
L34260.1 - 15 - 16 1880 86

View Full Product Details