The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281980.1 | NP_001268909.1 | 7 - 8 | 610 | 86 |
NM_001281981.1 | NP_001268910.1 | 8 - 9 | 651 | 86 | |
NM_008511.3 | NP_032537.3 | 7 - 8 | 882 | 86 | |
GenBank mRNA | AK040180.1 | - | 7 - 8 | 883 | 86 |
AK050646.1 | - | 7 - 8 | 1320 | 86 | |
AK079932.1 | - | 7 - 8 | 840 | 86 | |
AK153823.1 | - | 7 - 8 | 880 | 86 | |
AK160009.1 | - | 7 - 8 | 842 | 86 | |
AK166635.1 | - | 5 - 6 | 331 | 86 | |
AK168775.1 | - | 5 - 6 | 308 | 86 | |
AK169578.1 | - | 8 - 9 | 635 | 86 | |
AY753324.1 | - | 6 - 7 | 445 | 86 | |
BC052909.1 | - | 7 - 8 | 818 | 86 | |
U10484.1 | - | 7 - 8 | 588 | 86 |