The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011830.3 | NP_035960.2 | 9 - 10 | 1046 | 105 |
GenBank mRNA | AK002676.1 | - | 9 - 10 | 1078 | 105 |
AK011777.1 | - | 9 - 10 | 1073 | 105 | |
AK143505.1 | - | 9 - 10 | 1087 | 105 | |
AK146362.1 | - | 9 - 10 | 1112 | 105 | |
AK150713.1 | - | 9 - 10 | 1082 | 105 | |
AK151233.1 | - | 9 - 10 | 1082 | 105 | |
AK151706.1 | - | 9 - 10 | 1105 | 105 | |
AK152691.1 | - | 9 - 10 | 1079 | 105 | |
AK159822.1 | - | 9 - 10 | 1079 | 105 | |
AK160569.1 | - | 9 - 10 | 1079 | 105 | |
AK166667.1 | - | 9 - 10 | 1081 | 105 | |
AK169129.1 | - | 9 - 10 | 1112 | 105 | |
AK169788.1 | - | 9 - 10 | 1112 | 105 | |
BC052671.1 | - | 9 - 10 | 1067 | 105 | |
M33934.1 | - | 9 - 10 | 1079 | 105 | |
M98333.1 | - | 9 - 10 | 1013 | 105 | |
U89404.1 | - | 1 - 2 | 113 | 105 |