The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018739.2 | NP_061209.1 | 4 - 5 | 535 | 113 |
GenBank mRNA | AK146136.1 | - | 4 - 5 | 516 | 113 |
AK168391.1 | - | 4 - 5 | 528 | 113 | |
BC028484.1 | - | 2 - 3 | 113 | ||
BC048480.1 | - | 4 - 5 | 393 | 113 | |
BC115549.1 | - | 4 - 5 | 495 | 113 | |
BC115550.1 | - | 4 - 5 | 495 | 113 | |
D78255.1 | - | 4 - 5 | 514 | 113 |