Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arid3b
Entrez Gene ID
56380
Gene Name
AT rich interactive domain 3B (BRIGHT-like)
Gene Aliases
Bdp, Dri2
Chromosome Location
Chr.9: 57790353 - 57836847 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019689.2 NP_062663.1 4 - 5 955 73
GenBank mRNA AF116847.1 - 4 - 5 955 73
AK084952.1 - 2 - 3 267 73
AK139455.1 - 6 - 7 1137 73
AK148073.1 - 5 - 6 1052 73
BC145330.1 - 4 - 5 1019 73
BC150755.1 - 4 - 5 949 73

View Full Product Details