The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023134.4 | NP_075623.2 | 4 - 5 | 390 | 61 |
GenBank mRNA | AK004620.1 | - | 4 - 5 | 391 | 61 |
AK004788.1 | - | 4 - 5 | 391 | 61 | |
AK011333.1 | - | 3 - 4 | 364 | 61 | |
AK075702.1 | - | 4 - 5 | 394 | 61 |