The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285812.1 | NP_001272741.1 | 2 - 3 | 322 | 60 |
NM_001285814.1 | NP_001272743.1 | 3 - 4 | 387 | 60 | |
NM_001285815.1 | NP_001272744.1 | 3 - 4 | 405 | 60 | |
NM_026837.3 | NP_081113.1 | 2 - 3 | 301 | 60 | |
NM_029589.1 | NP_083865.1 | 3 - 4 | 223 | 60 | |
GenBank mRNA | AK004164.1 | - | 2 - 3 | 264 | 60 |
AK005104.1 | - | 2 - 3 | 208 | 60 | |
AK005885.1 | - | 3 - 4 | 227 | 60 | |
BC019937.1 | - | 2 - 3 | 227 | 60 | |
BC020121.1 | - | 2 - 3 | 231 | 60 | |
BC039805.1 | - | 1 - 2 | 606 | 60 |