The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138649.1 | NP_619590.1 | 1 - 2 | 212 | 84 |
GenBank mRNA | AB069667.1 | - | 1 - 2 | 212 | 84 |