The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008537.4 | NP_032563.2 | 3 - 4 | 606 | 116 |
GenBank mRNA | AK002401.1 | - | 3 - 4 | 574 | 116 |
AK048249.1 | - | 3 - 4 | 607 | 116 | |
AK160586.1 | - | 3 - 4 | 609 | 116 | |
BC015825.1 | - | 3 - 4 | 534 | 116 | |
U89906.1 | - | 3 - 4 | 514 | 116 |