Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adh7
Entrez Gene ID
11529
Gene Name
alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide
Gene Aliases
AI325182, Adh-3, Adh-3e, Adh-3t, Adh3, Adh3-e, Adh3-t, Adh4, Adt-1
Chromosome Location
Chr.3: 138198053 - 138232042 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009626.4 NP_033756.2 6 - 7 949 109
GenBank mRNA AF331802.1 - 6 - 7 947 109
AF331803.1 - 6 - 7 947 109
AK009588.2 - 6 - 7 916 109
AK009600.1 - 6 - 7 962 109
AK144795.1 - - 900 109
BC047267.1 - 6 - 7 878 109
U20257.1 - 6 - 7 840 109

View Full Product Details