The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025717.3 | NP_079993.2 | 1 - 2 | 136 | 101 |
GenBank mRNA | AK006447.1 | - | 1 - 2 | 119 | 101 |
AK014823.1 | - | 1 - 2 | 60 | 101 | |
AK019871.2 | - | 1 - 2 | 94 | 101 | |
AK032852.1 | - | 1 - 2 | 96 | 101 | |
AK142116.1 | - | 1 - 2 | 91 | 101 | |
AK160796.1 | - | 1 - 2 | 81 | 101 | |
BF782018.1 | - | - | 152 | 101 |