The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177576.1 | NP_001171047.1 | 4 - 5 | 418 | 100 |
NM_026646.3 | NP_080922.1 | 4 - 5 | 660 | 100 | |
GenBank mRNA | AK010193.1 | - | 4 - 5 | 406 | 100 |
AK018760.1 | - | 4 - 5 | 424 | 100 | |
AK046874.1 | - | 4 - 5 | 405 | 100 | |
AK048389.1 | - | 4 - 5 | 389 | 100 | |
AK049313.1 | - | 4 - 5 | 389 | 100 | |
AK155276.1 | - | 4 - 5 | 412 | 100 | |
AK162400.1 | - | 4 - 5 | 371 | 100 | |
BC037949.1 | - | 4 - 5 | 417 | 100 | |
BC050887.1 | - | 4 - 5 | 613 | 100 |