Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hspa12b
Entrez Gene ID
72630
Gene Name
heat shock protein 12B
Gene Aliases
2700081N06Rik
Chromosome Location
Chr.2: 131127412 - 131145985 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028306.3 NP_082582.1 4 - 5 389 91
GenBank mRNA AK012548.1 - 4 - 5 362 91
AY196790.1 - 3 - 4 269 91
BC011103.1 - 4 - 5 389 91

View Full Product Details