Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hspa12b
Entrez Gene ID
72630
Gene Name
heat shock protein 12B
Gene Aliases
2700081N06Rik
Chromosome Location
Chr.2: 131127412 - 131145985 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028306.3 NP_082582.1 6 - 7 676 60
GenBank mRNA AK012548.1 - 6 - 7 649 60
AY196790.1 - 5 - 6 556 60
BC011103.1 - 6 - 7 676 60

View Full Product Details