Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx16
Entrez Gene ID
74718
Gene Name
sorting nexin 16
Gene Aliases
4930522N22Rik, AI117577, AI747675
Chromosome Location
Chr.3: 10417817 - 10440131 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127191.2 NP_001120663.1 3 - 4 767 81
NM_029068.4 NP_083344.3 3 - 4 767 81
GenBank mRNA AK015872.1 - 3 - 4 728 81
AK032055.1 - 3 - 4 724 81
AK166422.1 - 3 - 4 741 81
BC019424.1 - 2 - 3 532 81
BC070431.1 - - 710 81

View Full Product Details