Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdyl2
Entrez Gene ID
75796
Gene Name
chromodomain protein, Y chromosome-like 2
Gene Aliases
1700029M19Rik, 4930453I21Rik, Gm20194
Chromosome Location
Chr.8: 116568724 - 116732991 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029441.3 NP_083717.1 3 - 4 991 58
GenBank mRNA AK006509.1 - 2 - 3 286 58
AK015452.1 - 3 - 4 845 58
AK142084.1 - 3 - 4 881 58
AY273799.1 - 3 - 4 840 58
BC058956.1 - 3 - 4 991 58

View Full Product Details