The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133981.2 | NP_598742.1 | 6 - 7 | 858 | 63 |
NR_045601.1 | 5 - 6 | 787 | 63 | ||
GenBank mRNA | AK050335.1 | - | 6 - 7 | 779 | 63 |
AK054293.1 | - | 6 - 7 | 764 | 63 | |
AK086674.1 | - | 6 - 7 | 798 | 63 | |
AK149592.1 | - | 5 - 6 | 709 | 63 | |
AK212096.1 | - | 4 - 5 | 365 | 63 | |
BC021791.1 | - | 6 - 7 | 753 | 63 |