The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109764.1 | NP_001103234.1 | 6 - 7 | 938 | 84 |
NM_009819.2 | NP_033949.2 | 5 - 6 | 836 | 84 | |
NM_145732.2 | NP_663785.2 | 6 - 7 | 938 | 84 | |
GenBank mRNA | AK035033.1 | - | 5 - 6 | 732 | 84 |
AK158916.1 | - | 6 - 7 | 971 | 84 | |
BC051995.1 | - | 5 - 6 | 717 | 84 | |
BC079648.1 | - | 6 - 7 | 855 | 84 | |
D25281.1 | - | 6 - 7 | 865 | 84 | |
D25282.1 | - | 5 - 6 | 765 | 84 |