The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013505.3 | NP_038533.1 | 1 - 2 | 324 | 53 |
GenBank mRNA | AK028604.1 | - | 1 - 2 | 329 | 53 |
AK028636.1 | - | 1 - 2 | 325 | 53 | |
AK030333.1 | - | 1 - 2 | 325 | 53 | |
AK048547.1 | - | 1 - 2 | 325 | 53 | |
AK172168.1 | - | 1 - 2 | 159 | 53 | |
BC057867.1 | - | 1 - 2 | 104 | 53 | |
L33779.1 | - | 1 - 2 | 123 | 53 |