The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053208.4 | NP_444438.2 | 2 - 3 | 1227 | 66 |
GenBank mRNA | AF340231.1 | - | 1 - 2 | 969 | 66 |
AF453879.1 | - | 2 - 3 | 1149 | 66 | |
AJ310547.1 | - | 2 - 3 | 1198 | 66 | |
AK075582.1 | - | 2 - 3 | 1179 | 66 | |
AK152056.1 | - | 2 - 3 | 1188 | 66 | |
AK152223.1 | - | 2 - 3 | 1185 | 66 | |
AK159206.1 | - | 2 - 3 | 1180 | 66 | |
AK166661.1 | - | 3 - 4 | 1093 | 66 | |
AK167477.1 | - | 2 - 3 | 1180 | 66 | |
BC023299.1 | - | 2 - 3 | 1161 | 66 | |
BC051436.1 | - | 1 - 2 | 945 | 66 | |
BC086764.1 | - | 2 - 3 | 1166 | 66 |