The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109973.2 | NP_001103443.1 | 3 - 4 | 644 | 84 |
NM_001276390.1 | NP_001263319.1 | 2 - 3 | 526 | 84 | |
NM_001276391.1 | NP_001263320.1 | 4 - 5 | 416 | 84 | |
NM_010783.3 | NP_034913.2 | 3 - 4 | 312 | 84 | |
GenBank mRNA | BC006018.1 | - | 2 - 3 | 279 | 84 |
BC010259.1 | - | 3 - 4 | 484 | 84 | |
BC083058.1 | - | 3 - 4 | 417 | 84 | |
BC085233.1 | - | 3 - 4 | 644 | 84 | |
CX564914.1 | - | 3 - 4 | 383 | 84 | |
U57820.1 | - | 2 - 3 | 284 | 84 | |
U57821.1 | - | 3 - 4 | 282 | 84 | |
U57822.1 | - | 2 - 3 | 464 | 84 |