The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144793.1 | NP_659042.1 | 3 - 4 | 379 | 59 |
GenBank mRNA | AK078385.1 | - | 3 - 4 | 395 | 59 |
BC010801.1 | - | 3 - 4 | 379 | 59 |