Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sin3b
Entrez Gene ID
20467
Gene Name
transcriptional regulator, SIN3B (yeast)
Gene Aliases
2810430C10Rik
Chromosome Location
Chr.8: 72723270 - 72758203 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113248.2 NP_001106719.1 2 - 3 248 73
NM_009188.4 NP_033214.2 2 - 3 248 73
GenBank mRNA AF038848.1 - 2 - 3 228 73
AK146847.1 - 2 - 3 231 73
AK159076.1 - 2 - 3 231 73
AK165592.1 - 2 - 3 233 73
AK169433.1 - 2 - 3 233 73
AK173005.1 - 2 - 3 217 73
BC020049.1 - 2 - 3 212 73
BC021160.1 - 2 - 3 231 73
BC096367.1 - 2 - 3 216 73
BC129859.1 - 2 - 3 220 73
L38622.2 - 2 - 3 220 73

View Full Product Details