Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gramd1b
Entrez Gene ID
235283
Gene Name
GRAM domain containing 1B
Gene Aliases
3222402H23, A930008A22Rik, AI593249, mKIAA1201
Chromosome Location
Chr.9: 40293233 - 40531499 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172768.1 NP_766356.1 13 - 14 1874 68
GenBank mRNA AK028277.1 - 14 - 15 1769 68
AK033156.1 - 13 - 14 1874 68
AK122465.1 - 12 - 13 1834 68
BC070451.1 - 12 - 13 1708 68

View Full Product Details