The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172768.1 | NP_766356.1 | 19 - 20 | 2640 | 92 |
GenBank mRNA | AK028277.1 | - | 20 - 21 | 2535 | 92 |
AK033156.1 | - | 19 - 20 | 2640 | 92 | |
AK122465.1 | - | 19 - 20 | 2612 | 92 | |
BC030911.1 | - | 10 - 11 | 1384 | 92 | |
BC070451.1 | - | 19 - 20 | 2486 | 92 |