Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vwa2
Entrez Gene ID
240675
Gene Name
von Willebrand factor A domain containing 2
Gene Aliases
Amaco
Chromosome Location
Chr.19: 56874416 - 56912078 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172840.2 NP_766428.2 7 - 8 869 74
GenBank mRNA AJ536329.1 - 7 - 8 869 74
AK029297.1 - 7 - 8 816 74
AK133486.1 - 7 - 8 776 74
AK160971.1 - 7 - 8 875 74
BC115868.1 - - 818 74
BC116636.1 - 7 - 8 819 74

View Full Product Details