The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175300.4 | NP_780509.2 | 12 - 13 | 2352 | 67 |
GenBank mRNA | AK033784.1 | - | 12 - 13 | 2320 | 67 |
AK129351.1 | - | 12 - 13 | 2305 | 67 | |
AK154465.1 | - | 12 - 13 | 2306 | 67 | |
AK170676.1 | - | 12 - 13 | 2318 | 67 | |
AK218217.1 | - | 1 - 2 | 96 | 67 | |
BC027351.1 | - | 12 - 13 | 2300 | 67 | |
BC038267.1 | - | 11 - 12 | 1663 | 67 | |
BC065421.1 | - | 3 - 4 | 465 | 67 | |
BC096381.1 | - | 11 - 12 | 2545 | 67 | |
BC137583.1 | - | 12 - 13 | 2320 | 67 |