The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175434.3 | NP_780643.2 | 3 - 4 | 824 | 92 |
GenBank mRNA | AK046359.1 | - | 3 - 4 | 804 | 92 |
AK083105.1 | - | 2 - 3 | 579 | 92 | |
BC115859.1 | - | 3 - 4 | 670 | 92 | |
BC115965.1 | - | 3 - 4 | 670 | 92 |