Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aifm2
Entrez Gene ID
71361
Gene Name
apoptosis-inducing factor, mitochondrion-associated 2
Gene Aliases
5430437E11Rik, Amid, D730001I10Rik, PRG3
Chromosome Location
Chr.10: 61715263 - 61739260 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039194.3 NP_001034283.1 8 - 9 1080 57
NM_001284300.1 NP_001271229.1 8 - 9 1080 57
NM_178058.4 NP_835159.1 8 - 9 1080 57
GenBank mRNA AK017403.1 - 8 - 9 1077 57
AK085656.1 - 8 - 9 1032 57
AK147741.1 - 8 - 9 1057 57
AK155240.1 - 8 - 9 1020 57
AK196892.1 - 1 - 2 213 57

View Full Product Details