Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nlrx1
Entrez Gene ID
270151
Gene Name
NLR family member X1
Gene Aliases
BC034204, NOD9
Chromosome Location
Chr.9: 44252713 - 44268599 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163742.1 NP_001157214.1 4 - 5 381 55
NM_001163743.1 NP_001157215.1 4 - 5 482 55
NM_178420.3 NP_848507.2 4 - 5 470 55
GenBank mRNA AK089260.1 - 4 - 5 474 55
AK141296.1 - 4 - 5 451 55
AK145780.1 - 4 - 5 491 55
AK145988.1 - 4 - 5 471 55
AK166689.1 - 4 - 5 308 55
AK170088.1 - 4 - 5 425 55
BC034204.1 - 3 - 4 264 55
BC050054.1 - 4 - 5 377 55

View Full Product Details