Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
D2hgdh
Entrez Gene ID
98314
Gene Name
D-2-hydroxyglutarate dehydrogenase
Gene Aliases
AA408776, AA408778, AI325464
Chromosome Location
Chr.1: 93824808 - 93852358 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178882.3 NP_849213.2 7 - 8 1138 61
GenBank mRNA AK088200.1 - - 1085 61
AK088554.1 - - 1085 61
AK141691.1 - 7 - 8 1368 61
AK170226.1 - - 1139 61
BC023277.1 - - 1106 61
BC117794.2 - 7 - 8 1076 61

View Full Product Details