The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177275.4 | NP_796249.1 | 1 - 1 | 950 | 100 |
GenBank mRNA | AB167510.1 | - | 1 - 1 | 898 | 100 |
AK088051.1 | - | 1 - 1 | 951 | 100 | |
AY237004.1 | - | 1 - 1 | 729 | 100 |