Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc42
Entrez Gene ID
276920
Gene Name
coiled-coil domain containing 42
Gene Aliases
A530001H01Rik
Chromosome Location
Chr.11: 68587037 - 68597954 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177779.3 NP_808447.2 5 - 6 889 59
GenBank mRNA AK079909.1 - 1 - 2 318 59
BC138437.1 - 1 - 2 316 59
BC138438.1 - 1 - 2 316 59

View Full Product Details