The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009275.4 | NP_033301.1 | 1 - 2 | 151 | 125 |
GenBank mRNA | AK004854.1 | - | 1 - 2 | 184 | 125 |
AK007859.1 | - | 1 - 2 | 157 | 125 | |
AK008383.1 | - | 1 - 2 | 194 | 125 | |
AK015469.1 | - | 1 - 2 | 216 | 125 | |
AK030437.1 | - | 1 - 2 | 152 | 125 | |
AK075583.1 | - | 1 - 2 | 154 | 125 | |
AK160600.1 | - | 1 - 2 | 217 | 125 | |
AK167039.1 | - | 1 - 2 | 157 | 125 | |
BC003798.1 | - | 1 - 2 | 172 | 125 | |
U17343.1 | - | 1 - 2 | 165 | 125 |