Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Igf2bp2
Entrez Gene ID
319765
Gene Name
insulin-like growth factor 2 mRNA binding protein 2
Gene Aliases
C330012H03Rik, IMP-2, Imp2, Neilsen
Chromosome Location
Chr.16: 22059009 - 22163299 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183029.2 NP_898850.2 14 - 15 1910 76
GenBank mRNA AK044984.1 - 13 - 14 1426 76
AK049196.1 - 14 - 15 1606 76
AK077404.1 - 14 - 15 1591 76
AK159654.1 - 14 - 15 1643 76
AK170531.1 - 13 - 14 1414 76
AY531659.1 - 14 - 15 1574 76
BC023758.1 - 14 - 15 1608 76
BC054552.1 - 13 - 14 3010 76
BU708344.1 - 4 - 5 355 76

View Full Product Details