Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vamp3
Entrez Gene ID
22319
Gene Name
vesicle-associated membrane protein 3
Gene Aliases
D130027G05Rik, VAMP-3, ceb
Chromosome Location
Chr.4: 151047305 - 151057953 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009498.4 NP_033524.1 4 - 5 375 125
GenBank mRNA AK051272.1 - 4 - 5 376 125
AK051691.1 - 4 - 5 370 125
AK165947.1 - 4 - 5 324 125
AK168413.1 - 4 - 5 364 125
AK169056.1 - 4 - 5 373 125
AK169087.1 - 4 - 5 367 125
BC060045.1 - 4 - 5 350 125
U60961.1 - 4 - 5 317 125

View Full Product Details