The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018798.2 | NP_061268.2 | 1 - 1 | 1862 | 143 |
GenBank mRNA | AF177346.1 | - | 1 - 1 | 1863 | 143 |
AK076159.1 | - | 1 - 1 | 1866 | 143 | |
BC021824.1 | - | 1 - 1 | 1244 | 143 | |
BC053022.1 | - | 1 - 1 | 1862 | 143 |