Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wnk4
Entrez Gene ID
69847
Gene Name
WNK lysine deficient protein kinase 4
Gene Aliases
2010002J11Rik, Pha2b, Prkwnk4
Chromosome Location
Chr.11: 101260537 - 101277409 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175638.3 NP_783569.1 10 - 11 2130 130
GenBank mRNA AY184228.1 - 10 - 11 2099 130
AY187027.1 - 10 - 11 2055 130
AY260059.1 - 10 - 11 2093 130
BC026679.1 - 3 - 4 329 130
BC037114.1 - 10 - 11 2090 130
BC043677.1 - 10 - 11 1503 130
BC096453.1 - 10 - 11 2086 130

View Full Product Details