Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tub
Entrez Gene ID
22141
Gene Name
tubby candidate gene
Gene Aliases
rd5, tub
Chromosome Location
Chr.7: 109010880 - 109034459 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021885.4 NP_068685.1 9 - 10 1331 101
GenBank mRNA AK147700.1 - 9 - 10 1386 101
BC096554.1 - 9 - 10 1312 101
U52433.1 - 9 - 10 1319 101
U52824.1 - 10 - 11 1464 101
U54643.1 - 9 - 10 1249 101

View Full Product Details