Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tub
Entrez Gene ID
22141
Gene Name
tubby candidate gene
Gene Aliases
rd5, tub
Chromosome Location
Chr.7: 109010880 - 109034459 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021885.4 NP_068685.1 8 - 9 1216 80
GenBank mRNA AK147700.1 - 8 - 9 1271 80
BC096554.1 - 8 - 9 1197 80
U52433.1 - 8 - 9 1204 80
U54643.1 - 8 - 9 1134 80

View Full Product Details