Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Keap1
Entrez Gene ID
50868
Gene Name
kelch-like ECH-associated protein 1
Gene Aliases
INRF2, mKIAA0132
Chromosome Location
Chr.9: 21229730 - 21239332 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110305.1 NP_001103775.1 3 - 4 1917 80
NM_001110306.1 NP_001103776.1 3 - 4 1802 80
NM_001110307.1 NP_001103777.1 3 - 4 1634 80
NM_016679.4 NP_057888.1 2 - 3 2953 80
GenBank mRNA AB020063.1 - 3 - 4 1406 80
AK004738.1 - 3 - 4 1634 80
AK039549.1 - 3 - 4 1573 80
AK046178.1 - 3 - 4 1518 80
AK076234.1 - 2 - 3 2439 80
AK129061.1 - 2 - 3 1388 80
AK136319.1 - 1 - 2 638 80
AK150437.1 - 3 - 4 1538 80
AK150485.1 - 3 - 4 1405 80
AK152142.1 - 3 - 4 1538 80
AK154430.1 - 3 - 4 1449 80
AK155507.1 - 3 - 4 1449 80
AK159858.1 - 3 - 4 1403 80
AK183261.1 - 1 - 2 80
BC055732.1 - 2 - 3 1391 80

View Full Product Details