Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mterf3
Entrez Gene ID
66410
Gene Name
mitochondrial transcription termination factor 3
Gene Aliases
2410017I18Rik, Mterfd1
Chromosome Location
Chr.13: 66905456 - 66933072 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025547.3 NP_079823.2 5 - 6 968 119
GenBank mRNA AK002299.1 - 5 - 6 909 119
AK010525.1 - 5 - 6 963 119
AK011934.1 - 5 - 6 956 119
BC025173.1 - 5 - 6 937 119
BC085282.1 - 5 - 6 938 119

View Full Product Details