Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tlr8
Entrez Gene ID
170744
Gene Name
toll-like receptor 8
Gene Aliases
-
Chromosome Location
Chr.X: 167241123 - 167264329 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY035890.1 - 1 - 2 66 96

View Full Product Details