The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163024.1 | NP_001156496.1 | 31 - 32 | 5052 | 113 |
NM_001163025.1 | NP_001156497.1 | 29 - 30 | 5034 | 113 | |
NM_153395.2 | NP_700444.2 | 29 - 30 | 5031 | 113 | |
GenBank mRNA | AB014059.1 | - | 29 - 30 | 4553 | 113 |
AK011826.1 | - | 8 - 9 | 1821 | 113 | |
AK012182.2 | - | 9 - 10 | 1970 | 113 | |
AK122425.1 | - | 31 - 32 | 5052 | 113 | |
AK162859.1 | - | 6 - 7 | 1525 | 113 | |
BC048406.1 | - | 18 - 19 | 2828 | 113 | |
BC150943.1 | - | 29 - 30 | 4555 | 113 | |
CV561694.1 | - | 3 - 4 | 448 | 113 |