The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025896.2 | NP_080172.1 | 4 - 5 | 649 | 127 |
GenBank mRNA | AF525154.1 | - | 4 - 5 | 634 | 127 |
AK005474.1 | - | 4 - 5 | 650 | 127 | |
AK005567.1 | - | 4 - 5 | 647 | 127 | |
BC100334.1 | - | 4 - 5 | 586 | 127 |