Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Elane
Entrez Gene ID
50701
Gene Name
elastase, neutrophil expressed
Gene Aliases
Ela2, F430011M15Rik, NE
Chromosome Location
Chr.10: 79886312 - 79888216 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015779.2 NP_056594.2 2 - 3 243 54
GenBank mRNA AK143710.1 - 2 - 3 243 54
BC145800.1 - 2 - 3 308 54

View Full Product Details