Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rufy2
Entrez Gene ID
70432
Gene Name
RUN and FYVE domain-containing 2
Gene Aliases
2610111M19Rik, AI844453, Denn, LZ-FYVE, ZFYVE13
Chromosome Location
Chr.10: 62980223 - 63018742 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027425.3 NP_081701.2 4 - 5 503 82
GenBank mRNA AK011847.1 - 4 - 5 473 82
AK083205.1 - 4 - 5 488 82
AK173195.1 - 4 - 5 464 82
BC017645.1 - 4 - 5 465 82
BC038265.1 - 4 - 5 444 82
BC137736.1 - 4 - 5 452 82
BC137737.1 - 4 - 5 452 82

View Full Product Details