Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ifitm7
Entrez Gene ID
74482
Gene Name
interferon induced transmembrane protein 7
Gene Aliases
4933438K12Rik, Mil4
Chromosome Location
Chr.16: 13981479 - 13986867 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270718.1 NP_001257647.1 1 - 2 236 78
GenBank mRNA AK076846.1 - 1 - 2 236 78

View Full Product Details